Nicotinamide hydroiodide ≥98%

Supplier: Matrix Scientific

085051-1G 085051-10G
102844-122EA 200 USD
102844-122 102844-124
Nicotinamide hydroiodide ≥98%
Nicotinamide hydroiodide

Hazard: Irritant

Formula: C₆H₇IN₂O
MDL Number: MFCD00072143
CAS Number: 3726-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR