2-(Aminosulfonyl)-N,N-dimethylnicotinamide ≥97%

Supplier: Matrix Scientific

Synonyms: N,N-Dimethyl-2-sulphamoylnicotinamide

085035-1G 085035-10G
102844-088EA 270.77 USD
102844-088 102844-090
2-(Aminosulfonyl)-N,N-dimethylnicotinamide ≥97%
2-(Aminosulfonyl)-N,N-dimethylnicotinamide

Hazard: Irritant

Formula: C₈H₁₁N₃O₃S
MW: 229.26 g/mol
MDL Number: MFCD03265507
CAS Number: 112006-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR