2-Methoxyisonicotinonitrile ≥97%

Supplier: Matrix Scientific

085022-1G 085022-10G
102844-054EA 69.23 USD
102844-054 102844-056
2-Methoxyisonicotinonitrile ≥97%
2-Methoxyisonicotinonitrile

Hazard: Irritant

Formula: C₇H₆N₂O
MDL Number: MFCD06659893
CAS Number: 72716-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR