3-Bromo-4-nitropyridine-N-oxide ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-4-nitropyridine 1-oxide

085017-1G 085017-10G
102844-042EA 69.23 USD
102844-042 102844-044
3-Bromo-4-nitropyridine-N-oxide ≥97%
3-Bromo-4-nitropyridine N-oxide

Hazard: Irritant

Formula: C₅H₃BrN₂O₃
MW: 219.00 g/mol
Melting Pt: 152-154°C
MDL Number: MFCD00085925
CAS Number: 1678-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR