1,2,3,4-Tetramethyl-5,6-dinitrobenzene ≥97%

Supplier: Matrix Scientific

085013-1G 085013-10G
102844-028EA 426.15 USD
102844-028 102844-030
1,2,3,4-Tetramethyl-5,6-dinitrobenzene ≥97%
1,2,3,4-Tetramethyl-5,6-dinitrobenzene

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₄
MDL Number: MFCD00052671
CAS Number: 18801-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR