2-Bromo-5-nitroanisole ≥97%

Supplier: Matrix Scientific

085007-1G 085007-10G
102844-006EA 86.15 USD
102844-006 102844-008
2-Bromo-5-nitroanisole ≥97%
2-Bromo-5-nitroanisole

Hazard: Irritant

Formula: C₇H₆BrNO₃
MDL Number: MFCD00041250
CAS Number: 77337-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR