2',4'-Dichloro-6'-methylacetanilide ≥97%
Supplier: Matrix Scientific
085003-1G
085003-10G
102843-994EA
270.77
USD
102843-994
102843-996
2',4'-Dichloro-6'-methylacetanilide ≥97%
2',4'-Dichloro-6'-methylacetanilide
Hazard: Irritant
Formula:
C₉H₉Cl₂NO MW: 218.08 g/mol |
CAS Number:
61655-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...