4'-Propylacetophenone oxime ≥97%

Supplier: Matrix Scientific

085002-1G 085002-10G
102843-990EA 241.54 USD
102843-990 102843-992
4'-Propylacetophenone oxime ≥97%
4'-Propylacetophenone oxime

Hazard: Irritant

Formula: C₁₁H₁₅NO
CAS Number: 64128-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR