4'-Propylacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Propylphenyl)ethanone, p-Propylacetophenone

085001-1G 085001-10G
102843-986EA 72.31 USD
102843-986 102843-988
4'-Propylacetophenone ≥95%
4'-Propylacetophenone

Hazard: Irritant

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
MDL Number: MFCD00041359
CAS Number: 2932-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR