1-(4-Nitrophenylsulfonyl)pyrrolidine ≥97%

Supplier: Matrix Scientific

084999-1G 084999-10G
102843-976EA 298.46 USD
102843-976 102843-978
1-(4-Nitrophenylsulfonyl)pyrrolidine ≥97%
1-(4-Nitrophenylsulphonyl)pyrrolidine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₄S
MDL Number: MFCD00277216
CAS Number: 175278-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR