4-(4-Nitrophenylsulfonyl)morpholine ≥97%
Supplier: Matrix Scientific
084998-1G
084998-10G
102843-972EA
270.77
USD
102843-972
102843-974
4-(4-Nitrophenylsulfonyl)morpholine ≥97%
4-(4-Nitrophenylsulphonyl)morpholine
Hazard: Irritant
Formula:
C₁₀H₁₂N₂O₅S |
MDL Number:
MFCD00277217 CAS Number: 1024-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...