2,4-Dimethylbenzophenone ≥97%

Supplier: Matrix Scientific

084996-1G 084996-10G 084996-50G
102843-962EA 86.15 USD
102843-962 102843-964 102843-966
2,4-Dimethylbenzophenone ≥97%
2,4-Dimethylbenzophenone

Hazard: Irritant

Formula: C₁₅H₁₄O
MDL Number: MFCD00008537
CAS Number: 1140-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR