N1-[4-(Allyloxy)phenyl]acetamide ≥95%

Supplier: Matrix Scientific

084995-1G 084995-10G 084995-50G
102843-956EA 72.31 USD
102843-956 102843-958 102843-960
N1-[4-(Allyloxy)phenyl]acetamide ≥95%
N1-[4-(Allyloxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD00127763
CAS Number: 6622-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR