3,4-Dimethylbenzamide ≥97%

Supplier: Matrix Scientific

084994-1G 084994-10G 084994-50G
102843-950EA 56.92 USD
102843-950 102843-952 102843-954
3,4-Dimethylbenzamide ≥97%
3,4-Dimethylbenzamide

Hazard: Irritant

Formula: C₉H₁₁NO
MDL Number: MFCD00052960
CAS Number: 5580-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR