3,4-Dichlorobenzaldehyde oxime ≥97%
Supplier: Matrix Scientific
Synonyms:
3,4-Dichlorobenzaldoxime
084990-1G
084990-10G
084990-50G
102843-938EA
80
USD
102843-938
102843-940
102843-942
3,4-Dichlorobenzaldehyde oxime ≥97%
3,4-Dichlorobenzaldehyde oxime
Hazard: Irritant
Formula:
C₇H₅Cl₂NO |
MDL Number:
MFCD00017592 CAS Number: 5331-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...