1-(4-Chlorophenyl)-2-(4-methylphenyl)ethane-1,2-dione ≥97%

Supplier: Matrix Scientific

084988-1G 084988-10G
102843-932EA 298.46 USD
102843-932 102843-934
1-(4-Chlorophenyl)-2-(4-methylphenyl)ethane-1,2-dione ≥97%
1-(4-Chlorophenyl)-2-(4-methylphenyl)ethane-1,2-dione

Hazard: Irritant

Formula: C₁₅H₁₁ClO₂
MDL Number: MFCD00052718
CAS Number: 86508-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR