4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile ≥97%

Supplier: Matrix Scientific

084985-1G 084985-10G
102843-922EA 241.54 USD
102843-922 102843-924
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile ≥97%
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile

Hazard: Irritant

Formula: C₁₁H₈N₂S
MDL Number: MFCD01765038
CAS Number: 127406-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR