2-(4-Isobutylphenyl)propanohydrazide ≥95%

Supplier: Matrix Scientific

084984-1G 084984-10G
102843-918EA 298.46 USD
102843-918 102843-920
2-(4-Isobutylphenyl)propanohydrazide ≥95%
2-(4-Isobutylphenyl)propanohydrazide

Hazard: Irritant

Formula: C₁₃H₂₀N₂O
MW: 220.32 g/mol
MDL Number: MFCD00711879
CAS Number: 127222-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR