Toluene-2-sulfonamide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methylbenzene-1-sulphonamide, 2-Methylbenzenesulphonamide, o-Toluenesulphonamide

084982-10G 084982-50G
102843-912EA 90.77 USD
102843-912 102843-914
Toluene-2-sulfonamide ≥97%
Toluene-2-sulphonamide

Hazard: Irritant

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
MDL Number: MFCD00007934
CAS Number: 88-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR