2'-Hydroxy-5'-methylacetanilide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Acetamido-4-methylphenol

084980-1G 084980-10G 084980-50G
102843-902EA 101.54 USD
102843-902 102843-904 102843-906
2'-Hydroxy-5'-methylacetanilide ≥97%
2'-Hydroxy-5'-methylacetanilide

Hazard: Irritant

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00020183
CAS Number: 6375-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR