1,4-Bis(2,2,2-trifluoroethoxy)benzene ≥97%

Supplier: Matrix Scientific

084979-1G 084979-10G
102843-898EA 86.15 USD
102843-898 102843-900
1,4-Bis(2,2,2-trifluoroethoxy)benzene ≥97%
1,4-Bis(2,2,2-trifluoroethoxy)benzene

Hazard: Irritant

Formula: C₁₀H₈F₆O₂
MW: 274.1597 g/mol
MDL Number: MFCD00042272
CAS Number: 66300-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR