N-(2-Methoxyphenethyl)urea ≥97%

Supplier: Matrix Scientific

084977-1G 084977-10G
102843-894EA 270.77 USD
102843-894 102843-896
N-(2-Methoxyphenethyl)urea ≥97%
N-(2-Methoxyphenethyl)urea

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD00174101
CAS Number: 69226-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR