2-Nitrophenyl phenyl ether ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Nitrodiphenylether, 1-Nitro-2-phenoxybenzene, 2-Nitrophenylphenylether

084964-1G 084964-10G
102843-854EA 101.54 USD
102843-854 102843-856
2-Nitrophenyl phenyl ether ≥97%
2-Nitrophenyl phenyl ether

TCA listed: T
Hazard: Irritant

Formula: C₁₂H₉NO₃
Boiling Pt: 195-197°C
Melting Pt: 20°C
Density: 1.254/21°C
MDL Number: MFCD00035744
CAS Number: 2216-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR