4,6-Di(tert-butyl)benzene-1,3-diol ≥97%

Supplier: Matrix Scientific

084956-1G 084956-10G
102843-834EA 115.38 USD
102843-834 102843-836
4,6-Di(tert-butyl)benzene-1,3-diol ≥97%
4,6-Di(tert-butyl)benzene-1,3-diol

Hazard: Irritant

Formula: C₁₄H₂₂O₂
Melting Pt: 121…123 °C
MDL Number: MFCD00044513
CAS Number: 5374-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR