3-(Dimethylamino)propiophenone hydrochloride ≥97%
Supplier: Matrix Scientific
084940-1G
084940-10G
102843-782EA
44.62
USD
102843-782
102843-784
3-(Dimethylamino)propiophenone hydrochloride ≥97%
3-(Dimethylamino)propiophenone hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₆ClNO MW: 213.71 g/mol |
MDL Number:
MFCD00012481 CAS Number: 879-72-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...