1-[2-Nitrovinyl]-2-(trifluoromethyl)benzene ≥97%

Supplier: Matrix Scientific

084924-1G 084924-10G
102843-748EA 170.77 USD
102843-748 102843-750
1-[2-Nitrovinyl]-2-(trifluoromethyl)benzene ≥97%
1-[2-Nitrovinyl]-2-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₆F₃NO₂
MDL Number: MFCD00192358
CAS Number: 53960-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR