5-Methyl-4-phenyl-1,2,3-thiadiazole ≥97%

Supplier: Matrix Scientific

084918-1G 084918-10G
102843-734EA 156.92 USD
102843-734 102843-736
5-Methyl-4-phenyl-1,2,3-thiadiazole ≥97%
5-Methyl-4-phenyl-1,2,3-thiadiazole

Hazard: Irritant

Formula: C₉H₈N₂S
MDL Number: MFCD00100201
CAS Number: 64273-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR