1-Chloro-2,4-dinitrobenzene ≥97%

Supplier: Matrix Scientific

084915-10G 084915-50G
102843-726EA 69.23 USD
102843-726 102843-728
1-Chloro-2,4-dinitrobenzene ≥97%
1-Chloro-2,4-dinitrobenzene

Hazard: Toxic

Formula: C₆H₃ClN₂O₄
Melting Pt: 48…50 °C
MDL Number: MFCD00007075
CAS Number: 97-00-7
UN: 3441
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR