3,5-Bis(trifluoromethyl)benzamidine hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3,5-Bis(trifluoromethyl)benzene-1-carboximidamide hydrochloride, 3,5-Bis(trifluoromethyl)phenylamidine hydrochloride

084909-1G 084909-10G
102843-708EA 156.92 USD
102843-708 102843-710
3,5-Bis(trifluoromethyl)benzamidine hydrochloride ≥97%
3,5-Bis(trifluoromethyl)benzamidine hydrochloride

Hazard: Irritant

Formula: C₉H₇ClF₆N₂
MW: 292.61 g/mol
MDL Number: MFCD00276604
CAS Number: 97603-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR