1-(2-Fluoro-5-nitrophenyl)-2,5-dimethylpyrrole ≥97%

Supplier: Matrix Scientific

Synonyms: 2,5-Dimethyl-1-(2-fluoro-5-nitrophenyl)-1H-pyrrole

084901-1G 084901-10G
102843-686EA 184.62 USD
102843-686 102843-688
1-(2-Fluoro-5-nitrophenyl)-2,5-dimethylpyrrole ≥97%
1-(2-Fluoro-5-nitrophenyl)-2,5-dimethylpyrrole

Hazard: Irritant

Formula: C₁₂H₁₁FN₂O₂
MW: 234.23 g/mol
MDL Number: MFCD02180649
CAS Number: 465514-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR