2-({[(4-Methylphenyl)sulfonyl]oxy}amino)ethanamine ≥97%

Supplier: Matrix Scientific

084885-1G 084885-10G
102843-646EA 213.85 USD
102843-646 102843-648
2-({[(4-Methylphenyl)sulfonyl]oxy}amino)ethanamine ≥97%
2-({[(4-Methylphenyl)sulfonyl]oxy}amino)ethanamine

Hazard: Irritant

Formula: C₉H₁₄N₂O₃S
MW: 230.29 g/mol
MDL Number: MFCD00276588
CAS Number: 175205-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR