2-Ethoxy-4-methylphenol ≥97%

Supplier: Matrix Scientific

084883-1G 084883-10G
102843-640EA 127.69 USD
102843-640 102843-642
2-Ethoxy-4-methylphenol ≥97%
2-Ethoxy-4-methylphenol

TCA listed: T
Hazard: Irritant

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 110°C/10 Torr
Melting Pt: 34-35°C
MDL Number: MFCD00043697
CAS Number: 2563-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR