4,4'-Dimethoxydiphenylamine ≥97%
Supplier: Matrix Scientific
Synonyms:
Bis(4-methoxyphenyl)amine
084871-1G
084871-10G
102843-612EA
270.77
USD
102843-612
102843-614
4,4'-Dimethoxydiphenylamine ≥97%
4,4'-Dimethoxydiphenylamine
Hazard: Irritant
Formula:
C₁₄H₁₅NO₂ MW: 229.28 g/mol Melting Pt: 100…104 °C |
MDL Number:
MFCD00014895 CAS Number: 101-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...