2-Chloro-5-nitroanisole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2-methoxy-4-nitrobenzene

084852-1G 084852-10G
102843-560EA 72.31 USD
102843-560 102843-562
2-Chloro-5-nitroanisole ≥97%
2-Chloro-5-nitroanisole

Hazard: Irritant

Formula: C₇H₆ClNO₃
MDL Number: MFCD00079739
CAS Number: 1009-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR