4-[(Trifluoromethyl)thio]benzonitrile ≥97%

Supplier: Matrix Scientific

084849-1G 084849-10G
102843-550EA 166.15 USD
102843-550 102843-552
4-[(Trifluoromethyl)thio]benzonitrile ≥97%
4-[(Trifluoromethyl)thio]benzonitrile

Hazard: Irritant

Formula: C₈H₄F₃NS
MW: 203.19 g/mol
MDL Number: MFCD00039475
CAS Number: 332-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR