2-[(2-Methylphenoxy)methyl]oxirane ≥90%

Supplier: Matrix Scientific

Synonyms: 2-((o-Tolyloxy)methyl)oxirane

084846-1G 084846-10G
102843-542EA 156.92 USD
102843-542 102843-544
2-[(2-Methylphenoxy)methyl]oxirane ≥90%
2-[(2-Methylphenoxy)methyl]oxirane

TCA listed: T
Hazard: Irritant

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
Boiling Pt: 119-20°C
Density: 1.079
Flash Pt: 120°C
MDL Number: MFCD00037847
CAS Number: 2210-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR