4-Chloro-3-ethylphenol ≥97%

Supplier: Matrix Scientific

084844-1G 084844-10G
102843-536EA 184.62 USD
102843-536 102843-538
4-Chloro-3-ethylphenol ≥97%
4-Chloro-3-ethylphenol

Hazard: Irritant

Formula: C₈H₉ClO
MW: 156.61 g/mol
MDL Number: MFCD00010822
CAS Number: 14143-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR