2'-Chloro-4'-(4-chlorophenoxy)acetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[2-Chloro-4-(4-chlorophenoxy)phenyl]ethan-1-one, 4-Acetyl-3,4'-dichlorodiphenyl ether

084843-1G 084843-10G
102843-532EA 213.85 USD
102843-532 102843-534
2'-Chloro-4'-(4-chlorophenoxy)acetophenone ≥95%
2'-Chloro-4'-(4-chlorophenoxy)acetophenone

Hazard: Irritant

Formula: C₁₄H₁₀Cl₂O₂
MW: 281.1340 g/mol
MDL Number: MFCD00140226
CAS Number: 119851-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR