2,3-Difluoro-4-iodophenol ≥97%

Supplier: Matrix Scientific

084841-1G 084841-10G
102843-526EA 100 USD
102843-526 102843-528
2,3-Difluoro-4-iodophenol ≥97%
2,3-Difluoro-4-iodophenol

Hazard: Irritant

Formula: C₆H₃F₂IO
MDL Number: MFCD08741391
CAS Number: 144292-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR