3-Chlorothiobenzamide ≥97%

Supplier: Matrix Scientific

084821-1G 084821-10G
102843-480EA 101.54 USD
102843-480 102843-482
3-Chlorothiobenzamide ≥97%
3-Chlorothiobenzamide

Hazard: Irritant

Formula: C₇H₆ClNS
MDL Number: MFCD00276993
CAS Number: 2548-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR