4-Methoxy-3-nitrobenzamide ≥97%

Supplier: Matrix Scientific

084818-1G 084818-10G
102843-472EA 184.62 USD
102843-472 102843-474
4-Methoxy-3-nitrobenzamide ≥97%
4-Methoxy-3-nitrobenzamide

Hazard: Irritant

Formula: C₈H₈N₂O₃
MW: 180.17 g/mol
MDL Number: MFCD00017136
CAS Number: 10397-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR