3'-Chloro-4'-methylacetanilide ≥97%

Supplier: Matrix Scientific

Synonyms: N-(3-Chloro-4-methylphenyl)acetamide

084817-1G 084817-10G 084817-50G
102843-466EA 101.54 USD
102843-466 102843-468 102843-470
3'-Chloro-4'-methylacetanilide ≥97%
3'-Chloro-4'-methylacetanilide

Hazard: Irritant

Formula: C₉H₁₀ClNO
MW: 183.64 g/mol
MDL Number: MFCD00032582
CAS Number: 7149-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR