4-(2-Methoxyethyl)phenol ≥97%
Supplier: Matrix Scientific
Synonyms:
p-(2-Methoxyethyl) phenol
084810-1G
084810-10G
084810-50G
102843-434EA
72.31
USD
102843-434
102843-436
102843-438
4-(2-Methoxyethyl)phenol ≥97%
4-(2-Methoxyethyl)phenol
Hazard: Irritant
Formula:
C₉H₁₂O₂ MW: 152.19 g/mol |
MDL Number:
MFCD00017537 CAS Number: 56718-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...