4-(2-Fluoro-4-nitrophenyl)morpholine ≥97%
Supplier: Matrix Scientific
084809-1G
084809-10G
102843-430EA
184.62
USD
102843-430
102843-432
4-(2-Fluoro-4-nitrophenyl)morpholine ≥97%
4-(2-Fluoro-4-nitrophenyl)morpholine
Hazard: Irritant
Formula:
C₁₀H₁₁FN₂O₃ MW: 226.20 g/mol |
MDL Number:
MFCD03138381 CAS Number: 2689-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...