4-(4-Cyanophenyl)morpholine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Morpholinobenzonitrile, 4-(4-Morpholinyl)benzonitrile

084808-1G 084808-10G
102843-426EA 127.69 USD
102843-426 102843-428
4-(4-Cyanophenyl)morpholine ≥97%
4-(4-Cyanophenyl)morpholine

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MW: 188.23 g/mol
MDL Number: MFCD00100115
CAS Number: 10282-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR