4-(4-Methylphenylthio)benzophenone ≥97%
Supplier: Matrix Scientific
084805-1G
084805-10G
102843-418EA
72.31
USD
102843-418
102843-420
4-(4-Methylphenylthio)benzophenone ≥97%
4-(4-Methylphenylthio)benzophenone
Hazard: Irritant
Formula:
C₂₀H₁₆OS |
MDL Number:
MFCD00055651 CAS Number: 83846-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...