2-Methoxy-4-nitrophenol ≥97%

Supplier: Matrix Scientific

084792-1G 084792-10G
102843-390EA 72.31 USD
102843-390 102843-392
2-Methoxy-4-nitrophenol ≥97%
2-Methoxy-4-nitrophenol

Hazard: Irritant

Formula: C₇H₇NO₄
MDL Number: MFCD00012143
CAS Number: 3251-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR