(4-Fluorophenyl)(oxo)acetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Fluorobenzoyl cyanide

084791-1G 084791-10G
102843-386EA 327.69 USD
102843-386 102843-388
(4-Fluorophenyl)(oxo)acetonitrile ≥97%
(4-Fluorophenyl)(oxo)acetonitrile

Hazard: Irritant

Formula: C₈H₄FNO
MW: 149.13 g/mol
MDL Number: MFCD06658451
CAS Number: 658-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR