trans-4-Isopropyl-β-nitrostyrene ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Isopropyl-β-nitrostyrene
084790-1G
084790-10G
084790-50G
102843-380EA
127.69
USD
102843-380
102843-382
102843-384
trans-4-Isopropyl-β-nitrostyrene ≥97%
trans-4-Isopropyl-β-nitrostyrene
Hazard: Irritant
Formula:
C₁₁H₁₃NO₂ MW: 191.23 g/mol |
MDL Number:
MFCD00085100 CAS Number: 42139-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...