2-(2,6-Dichlorophenyl)ethanimidamide hydrochloride ≥97%

Supplier: Matrix Scientific

084780-1G 084780-10G
102843-360EA 143.08 USD
102843-360 102843-362
2-(2,6-Dichlorophenyl)ethanimidamide hydrochloride ≥97%
2-(2,6-Dichlorophenyl)ethanimidamide hydrochloride

Hazard: Irritant

Formula: C₈H₉Cl₃N₂
MDL Number: MFCD00053014
CAS Number: 175276-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR